missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-5-(trifluoromethyl)furan, 97%, Thermo Scientific™

Catalog No. MO00141CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO00141CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO00141CB Supplier Thermo Scientific Chemicals Supplier No. MO00141CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17515-77-4
Molecular Formula C6H4BrF3O
Molecular Weight (g/mol) 229.00
MDL Number MFCD03086219
InChI Key YNHVBNGRNNVEMD-UHFFFAOYSA-N
Synonym 2-bromomethyl-5-trifluoromethyl furan, 2-bromomethyl-5-trifluoromethyl-furan, furan, 2-bromomethyl-5-trifluoromethyl, pubchem13345, 5-trifluoromethyl furfuryl bromide, 2-bromomethyl-5-trifloromethyl furan, 5-bromomethyl-2-trifluoromethyl furan
PubChem CID 2794605
IUPAC Name 2-(bromomethyl)-5-(trifluoromethyl)furan
SMILES FC(F)(F)C1=CC=C(CBr)O1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 228.99
Percent Purity 97%
Chemical Name or Material 2-(Bromomethyl)-5-(trifluoromethyl)furan
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.