missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisonicotinic Acid 98.0+%, TCI America™

Catalog No. p-7122903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33681G 1 g
B33685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33681G Supplier TCI America Supplier No. B33681G

Chemical Identifiers

CAS 66572-56-3
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.007
MDL Number MFCD01646069
InChI Key YBTKGKVQEXAYEM-UHFFFAOYSA-N
Synonym 2-bromoisonicotinic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromo-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 2-bromo, 2-bromoisonicotic acid, 2-bromisonicotins, 2-bromopyridine-4-carboxylicacid, pubchem9211, acmc-209nvt
PubChem CID 222701
IUPAC Name 2-bromopyridine-4-carboxylic acid
SMILES C1=CN=C(C=C1C(=O)O)Br

Specifications

Melting Point 229°C
Color White-Yellow
Quantity 1 g
Formula Weight 202.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoisonicotinic Acid