missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisobutyryl Bromide 98.0+%, TCI America™

Catalog No. B0607500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0607500G 500 g
B060725G 25 g
B0607100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0607500G Supplier TCI America Supplier No. B0607500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20769-85-1
Molecular Formula C4H6Br2O
Molecular Weight (g/mol) 229.899
MDL Number MFCD00000122
InChI Key YOCIJWAHRAJQFT-UHFFFAOYSA-N
Synonym 2-bromo-2-methylpropionyl bromide, 2-bromoisobutyryl bromide, propanoyl bromide, 2-bromo-2-methyl, bibb, 2-bromo-2-methylpropionylbromide, unii-627f9n9dya, 2-bromoisobutyrylbromide, 2-bromo-2-methyl-propionyl bromide, alpha-bromoisobutyryl bromide, 2-bromo-2-methyl-propanoyl bromide
PubChem CID 88685
IUPAC Name 2-bromo-2-methylpropanoyl bromide
SMILES CC(C)(C(=O)Br)Br

Specifications

Color Yellow
Boiling Point 164°C
Quantity 500 g
UN Number 3265
Formula Weight 229.90
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromoisobutyryl Bromide
EINECSNumber (2)-3443
TSCA Yes