missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofuran (stabilized with CaO) 98.0+%, TCI America™

Catalog No. p-7123613
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45551G 1 g
B45555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45551G Supplier TCI America Supplier No. B45551G

Chemical Identifiers

CAS 584-12-3
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00216609
InChI Key OYMCMWPHMPODNK-UHFFFAOYSA-N
Synonym furan, 2-bromo, bromofuran, 2-bromo-furan, 2-furyl bromide, alpha-furyl bromide, 2-bromofurane, 5-bromofuran, pubchem6929, acmc-209m4d
PubChem CID 2776190
IUPAC Name 2-bromofuran
SMILES BrC1=CC=CO1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 1 g
UN Number 1993
Formula Weight 146.97
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromofuran (stabilized with CaO)
TSCA No
Recommended Storage Freezer