missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl Phenyl Sulfone 98.0+%, TCI America™

Catalog No. B23671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23671G Supplier TCI America Supplier No. B23671G

Chemical Identifiers

CAS 92022-50-9
Molecular Formula C13H11BrO2S
Molecular Weight (g/mol) 311.193
MDL Number MFCD06797069
InChI Key CQNGEPLMWCLSBR-UHFFFAOYSA-N
Synonym 1-(Benzenesulfonylmethyl)-2-bromobenzene, 1-Bromo-2-(phenylsulfonylmethyl)benzene
PubChem CID 11335905
IUPAC Name 1-(benzenesulfonylmethyl)-2-bromobenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)CC2=CC=CC=C2Br

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 311.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzyl Phenyl Sulfone