missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzyl Alcohol 99.0+%, TCI America™

Catalog No. B129425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B129425G 25 g
B1294100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B129425G Supplier TCI America Supplier No. B129425G

Chemical Identifiers

CAS 18982-54-2
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00004600
InChI Key IOWGHQGLUMEZKG-UHFFFAOYSA-N
Synonym 2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746
PubChem CID 72850
IUPAC Name (2-bromophenyl)methanol
SMILES OCC1=CC=CC=C1Br

Specifications

Melting Point 80°C
Color White
Quantity 25 g
Formula Weight 187.04
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzyl Alcohol