missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzothiazole 95.0+%, TCI America™

Catalog No. B37401G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37401G Supplier TCI America Supplier No. B37401G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2516-40-7
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD02681887
InChI Key DRLMMVPCYXFPEP-UHFFFAOYSA-N
Synonym 2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
PubChem CID 612040
IUPAC Name 2-bromo-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)Br

Specifications

Melting Point 41°C
Color White
Boiling Point 125°C
Quantity 1 g
Formula Weight 214.08
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzothiazole