missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzoic acid, 98%

Catalog No. AAA1360830
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1360830 250 g
AAA1360818 50 g
AAA136080B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1360830 Supplier Thermo Scientific Chemicals Supplier No. A1360830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-65-3 | C7H5BrO2 | 201.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-65-3
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00002402
InChI Key XRXMNWGCKISMOH-UHFFFAOYSA-N
Synonym o-bromobenzoic acid, benzoic acid, 2-bromo, bromobenzoic acid, benzoic acid, o-bromo, 2-bromo-benzoic acid, benzoic acid, bromo, unii-az789tzs4l, az789tzs4l, ortho-bromobenzoic acid, attercop-chm at111657
PubChem CID 6940
IUPAC Name 2-bromobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)Br

Specifications

Melting Point 146°C to 150°C
Density 1.93
Quantity 250 g
Beilstein 971266
Formula Weight 201.02
Percent Purity 98%
Chemical Name or Material 2-Bromobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-848-6
RTECSNumber DG4448035
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only