missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzenethiol 98.0+%, TCI America™

Catalog No. B23475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23475G 5 g
B234725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23475G Supplier TCI America Supplier No. B23475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6320-02-1
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004827
InChI Key YUQUNWNSQDULTI-UHFFFAOYSA-N
Synonym 2-bromothiophenol, o-bromothiophenol, o-bromobenzenethiol, 2-bromo thiophenol, benzenethiol, 2-bromo, benzenethiol, o-bromo, 2-bromobenzene-1-thiol, 2-bromo-benzenethiol, 2-bromo-thiophenol, 2-bromobenzene thiol
PubChem CID 80598
IUPAC Name 2-bromobenzenethiol
SMILES C1=CC=C(C(=C1)S)Br

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
UN Number 1760
Formula Weight 189.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromobenzenethiol
TSCA Yes
Recommended Storage Refrigerator