missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-N-Fmoc-L-phenylalanine, 95%

Catalog No. AAH6642406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6642406 5 g
AAH6642414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6642406 Supplier Thermo Scientific Chemicals Supplier No. H6642406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 220497-47-2 | C24H20BrNO4 | 466.331 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 220497-47-2
Molecular Formula C24H20BrNO4
Molecular Weight (g/mol) 466.331
MDL Number MFCD01311743
InChI Key GRJPAUULVKPBHU-QFIPXVFZSA-N
Synonym fmoc-l-2-bromophenylalanine, fmoc-phe 2-br-oh, fmoc-2-bromo-l-phenylalanine, s-n-fmoc-2-bromophenylalanine, fmoc-l-2-bromophe, fmoc-d-2-bromophenylalanine, fmoc-l-2-br-phe-oh, 2s-3-2-bromophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-bromophenyl propanoic acid, 2s-3-2-bromophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 2734447
IUPAC Name (2S)-3-(2-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C(=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)Br

Specifications

Color White
Quantity 5 g
Formula Weight 426.47
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Bromo-N-Fmoc-L-phenylalanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only