missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9-fluorenone 98.0+%, TCI America™

Catalog No. B31095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31095G 5 g
B310925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31095G Supplier TCI America Supplier No. B31095G

Chemical Identifiers

CAS 3096-56-8
Molecular Formula C13H7BrO
Molecular Weight (g/mol) 259.10
MDL Number MFCD00010789
InChI Key MTCARZDHUIEYMB-UHFFFAOYSA-N
Synonym 2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f
PubChem CID 725831
IUPAC Name 2-bromo-9H-fluoren-9-one
SMILES BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 147°C
Color Yellow
Quantity 5 g
Formula Weight 259.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9-fluorenone