missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9,9-dimethylfluorene, 98%

Catalog No. AAH6473222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6473222 100 g
AAH6473206 5 g
AAH6473214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6473222 Supplier Thermo Scientific Chemicals Supplier No. H6473222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28320-31-2 | C15H13Br | 273.173 g/mol

This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 28320-31-2
Molecular Formula C15H13Br
Molecular Weight (g/mol) 273.173
MDL Number MFCD09261272
InChI Key MBHPOBSZPYEADG-UHFFFAOYSA-N
Synonym 2-bromo-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-2-bromofluorene, 9h-fluorene, 2-bromo-9,9-dimethyl, 2-bromo-9,9-dimethyl-fluorene, 2-bromo-9,9-dimethylfluoren, ksc494q3r, 2-bromo-9,9-dimethyl fluorene, 9,9-dimethyl-2-bromofluorenone, 2-bromo-9,9-dimethylfluorene
PubChem CID 10945606
IUPAC Name 2-bromo-9,9-dimethylfluorene
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)C

Specifications

Melting Point 57°C to 62°C
Quantity 100 g
Beilstein 2524870
Solubility Information Slightly soluble in water.
Formula Weight 273.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-9,9-dimethylfluorene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only