missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-(trifluoromethyl)pyridine, 97%

Catalog No. AAH2574906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2574906 5 g
AAH2574903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2574906 Supplier Thermo Scientific Chemicals Supplier No. H2574906
Only null left

CAS: 189278-27-1 | C6H3BrF3N | 226.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 189278-27-1
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 226.00
MDL Number MFCD00153087
InChI Key DOWNSQADAFSSAR-UHFFFAOYSA-N
Synonym 2-bromo-6-trifluoromethyl pyridine, pyridine, 2-bromo-6-trifluoromethyl, 2-trifluoromethyl-6-bromopyridine, 2-bromo-6-trifluoromethyl-pyridine, 6-bromo-2-trifluoromethyl pyridine, 2-bromo-6-trifluromethylpyridine, 2-bromo-6-trifluormethyl-pyridine, pubchem9307, acmc-209esf, ksc495o4t
PubChem CID 2782902
IUPAC Name 2-bromo-6-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=NC(Br)=CC=C1

Specifications

Melting Point 48°C to 51°C
Boiling Point 78°C to 79°C (14 mmHg)
Quantity 5 g
Formula Weight 226
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only