missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methylpyridine 98.0+%, TCI America™

Catalog No. B21145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21145G 5 g
B211425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21145G Supplier TCI America Supplier No. B21145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5315-25-3 | C6H6BrN | 172.03 g/mol

Chemical Identifiers

CAS 5315-25-3
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00040743
InChI Key SOHDPICLICFSOP-UHFFFAOYSA-N
Synonym 2-bromo-6-picoline, 6-bromo-2-picoline, 6-bromo-2-methylpyridine, 2-bromo-6-methyl-pyridine, 2-bromo-6-methyl pyridine, 2-brom-6-methylpyridin, pyridine, 2-bromo-6-methyl, 2-bromo-6-methy pyridine, 6-bromopicoline, 6-methyl-2-bromopyridine
PubChem CID 79205
IUPAC Name 2-bromo-6-methylpyridine
SMILES CC1=CC=CC(Br)=N1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromo-6-methylpyridine