missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-methoxypyridine 97.0+%, TCI America™

Catalog No. B32605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32605G 5 g
B326025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32605G Supplier TCI America Supplier No. B32605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40473-07-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD00088345
InChI Key KMODISUYWZPVGV-UHFFFAOYSA-N
Synonym 2-bromo-6-methoxy-pyridine, pyridine, 2-bromo-6-methoxy, 6-bromo-2-methoxypyridine, 2-methoxy-6-bromo pyridine, buttpark 82\06-06, 2-brom-6-methoxypyridin, zlchem 781, pubchem9206, acmc-209jec, 2-bromo-6-methoxy-pyridin
PubChem CID 256810
IUPAC Name 2-bromo-6-methoxypyridine
SMILES COC1=CC=CC(Br)=N1

Specifications

Color Yellow
Boiling Point 91°C
Quantity 5 g
Formula Weight 188.02
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-6-methoxypyridine