missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluoroaniline 98.0+%, TCI America™

Catalog No. B48035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48035G 5 g
B480325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48035G Supplier TCI America Supplier No. B48035G
Only null left

Chemical Identifiers

CAS 65896-11-9
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.02
MDL Number MFCD01310982
InChI Key ALZFPYUPNVLVQM-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-6-fluoro, 2-bromo-6-fluoro-phenylamine, 2-bromo-6-fluorobenzenamine, 2-bromo-6-fluoro-aniline, 2-bromo-6-fluoro aniline, 2-bromo-6-fluorophenylamine, pubchem2923, acmc-209nsu, 2-bromo-6-flouroaniline, ksc493o8h
PubChem CID 2782940
IUPAC Name 2-bromo-6-fluoroaniline
SMILES NC1=C(F)C=CC=C1Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-6-fluoroaniline