missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-(trifluoromethyl)pyridine, 96%

Catalog No. AA4338106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4338106 5 g
AA4338103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4338106 Supplier Thermo Scientific Chemicals Supplier No. 04338106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50488-42-1 | C6H3BrF3N | 225.996 g/mol

2-Bromo-5-(trifluoromethyl)pyridine is a substrate used in a palladium-catalyzed α-arylation of a Refomatsky reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50488-42-1
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD00153086
InChI Key GSKMWMFOQQBVMI-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethyl pyridine, 6-bromo-3-trifluoromethylpyridine, 2-bromo-5-trifluoromethyl-pyridine, pyridine, 2-bromo-5-trifluoromethyl, 5-trifluoromethyl-2-bromopyridine, 2-brom-5-trifluormethyl pyridin, 2-bromo-5-trilfuoromethylpyridine, pubchem3005
PubChem CID 2736434
IUPAC Name 2-bromo-5-(trifluoromethyl)pyridine
SMILES C1=CC(=NC=C1C(F)(F)F)Br

Specifications

Melting Point 45°C to 47°C
Boiling Point 78°C to 81°C (30 mmHg)
Quantity 5 g
Solubility Information Soluble in methanol and ethanol.
Formula Weight 226
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material 2-Bromo-5- (trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only