missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-phenylthiophene 97.0+%, TCI America™

Numéro de catalogue. B36631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B36631G 1 g
B36635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B36631G Fournisseur TCI America Code fournisseur B36631G
Il en reste null

Identifiants chimiques

CAS 29488-24-2
Molecular Formula C10H7BrS
Molecular Weight (g/mol) 239.13
MDL Number MFCD00159575
InChI Key RVCTZBRMQZWVDA-UHFFFAOYSA-N
PubChem CID 2776313
IUPAC Name 2-bromo-5-phenylthiophene
SMILES C1=CC=C(C=C1)C2=CC=C(S2)Br

Spécifications

Melting Point 85°C
Color White-Yellow
Quantity 1 g
Synonym thiophene,2-bromo-5-phenyl, 2-bromo-5-phenyl-thiophene, 2-phenyl-5-bromothiophene, thiophene, 2-bromo-5-phenyl, 5-bromo-2-phenylthiophene, acmc-209h9c, 2-bromo-5-phenyl thiophene
Formula Weight 239.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-phenylthiophene