missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-nitrobenzotrifluoride 98.0+%, TCI America™
Supplier: TCI America B368625G
Specifications
| 2-Bromo-5-nitrobenzotrifluoride | |
| 44°C | |
| C7H3BrF3NO2 | |
| 25 g | |
| SXEQQBBOAMHOID-UHFFFAOYSA-N | |
| 1-bromo-4-nitro-2-(trifluoromethyl)benzene | |
| 136198 | |
| ≥98.0% (GC) |
| 367-67-9 | |
| 88°C | |
| MFCD00014707 | |
| 2-bromo-5-nitrobenzotrifluoride, 1-bromo-4-nitro-2-trifluoromethyl benzene, 5-nitro-2-bromobenzotrifluoride, 2-bromo-5-nitro-a,a,a-trifluorotoluene, 4-bromo-3-trifluoromethyl nitrobenzene, benzene, 1-bromo-4-nitro-2-trifluoromethyl, pubchem10394, 2-bromo-5-nirtobenzotrifluoride, 2-bromo-5-nitro-benzotrifluoride, attercop-chm at115179 | |
| [O-][N+](=O)C1=CC=C(Br)C(=C1)C(F)(F)F | |
| 270.01 | |
| 270.01 | |
| Crystalline Powder |
Chemical Identifiers
| 367-67-9 | |
| 270.01 | |
| SXEQQBBOAMHOID-UHFFFAOYSA-N | |
| 136198 | |
| [O-][N+](=O)C1=CC=C(Br)C(=C1)C(F)(F)F |
| C7H3BrF3NO2 | |
| MFCD00014707 | |
| 2-bromo-5-nitrobenzotrifluoride, 1-bromo-4-nitro-2-trifluoromethyl benzene, 5-nitro-2-bromobenzotrifluoride, 2-bromo-5-nitro-a,a,a-trifluorotoluene, 4-bromo-3-trifluoromethyl nitrobenzene, benzene, 1-bromo-4-nitro-2-trifluoromethyl, pubchem10394, 2-bromo-5-nirtobenzotrifluoride, 2-bromo-5-nitro-benzotrifluoride, attercop-chm at115179 | |
| 1-bromo-4-nitro-2-(trifluoromethyl)benzene |
Safety and Handling
TSCA : No