accessibility menu, dialog, popup

UserName

CAS RN 367-67-9

IUPAC Name: 1-bromo-4-nitro-2-(trifluoromethyl)benzene
Synonyms: 1-bromo-4-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 270.01
Molecular Formula: C7H3BrF3NO2
InChi Key: SXEQQBBOAMHOID-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)C(=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results