missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7123102
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36865G 5 g
B368625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36865G Supplier TCI America Supplier No. B36865G

Chemical Identifiers

CAS 367-67-9
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.01
MDL Number MFCD00014707
InChI Key SXEQQBBOAMHOID-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrobenzotrifluoride, 1-bromo-4-nitro-2-trifluoromethyl benzene, 5-nitro-2-bromobenzotrifluoride, 2-bromo-5-nitro-a,a,a-trifluorotoluene, 4-bromo-3-trifluoromethyl nitrobenzene, benzene, 1-bromo-4-nitro-2-trifluoromethyl, pubchem10394, 2-bromo-5-nirtobenzotrifluoride, 2-bromo-5-nitro-benzotrifluoride, attercop-chm at115179
PubChem CID 136198
IUPAC Name 1-bromo-4-nitro-2-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC=C(Br)C(=C1)C(F)(F)F

Specifications

Melting Point 44°C
Boiling Point 88°C
Quantity 5 g
Formula Weight 270.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-nitrobenzotrifluoride