missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals

Catalog No. AAH50608MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50608MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH50608MD Supplier Thermo Scientific Chemicals Supplier No. H50608MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 138642-47-4
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.046
MDL Number MFCD01861260
InChI Key ZQONVYONOASKIY-UHFFFAOYSA-N
Synonym benzonitrile,2-bromo-5-methoxy, 2-bromo-5-methoxybenzenecarbonitrile, 4-bromo-3-cyanoanisole, acmc-20ag98, 2-bromo-5-methoxy benzonitrile, 2-bromo-5-methoxybenzonitrile, 2-bromanyl-5-methoxy-benzenecarbonitrile
PubChem CID 21784096
IUPAC Name 2-bromo-5-methoxybenzonitrile
SMILES COC1=CC(=C(C=C1)Br)C#N

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 212.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-methoxybenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only