missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-iodotoluene 98.0+%, TCI America™

Catalog No. B30995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30995G 5 g
B309925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30995G Supplier TCI America Supplier No. B30995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 202865-85-8
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
MDL Number MFCD00070749
InChI Key QSQOGKONVJDRNH-UHFFFAOYSA-N
PubChem CID 2735574
IUPAC Name 1-bromo-4-iodo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)I)Br

Specifications

Color Yellow
Quantity 5 g
Synonym 2-bromo-5-iodotoluene, benzene,1-bromo-4-iodo-2-methyl, benzene, 1-bromo-4-iodo-2-methyl, pubchem3791, 2-bromo-5-iodo toluene, 2-bromo-5-iodo-toluene, 2-bromo-5-iodotoluene;, acmc-1cq3e, ksc543o8j, 1-bromo-2-methyl-4-iodobenzene
Formula Weight 296.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-iodotoluene