missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoropyridine 98.0+%, TCI America™

Catalog No. B27995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27995G 5 g
B279925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27995G Supplier TCI America Supplier No. B27995G

Chemical Identifiers

CAS 41404-58-4
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
InChI Key UODINHBLNPPDPD-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoro-pyridine, pyridine, 2-bromo-5-fluoro, pubchem1099, pubchem14324, 2-bromo5-fluoropyridine, acmc-1assy, 2-bromo-5-fluoropyridin, 2-bromo-5-fluorpyridine, 2-bromo-5-fluropyridine, 5-fluoro-2-bromopyridine
PubChem CID 2783171
IUPAC Name 2-bromo-5-fluoropyridine
SMILES C1=CC(=NC=C1F)Br

Specifications

Color White-Yellow
Boiling Point 83°C
Quantity 5 g
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Bromo-5-fluoropyridine