missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluoroaniline, 98%

Catalog No. AAB2490114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2490114 25 g
AAB2490106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2490114 Supplier Thermo Scientific Chemicals Supplier No. B2490114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1003-99-2 | C6H5BrFN | 190.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1003-99-2
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00070750
InChI Key FWTXFEKVHSFTDQ-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-5-fluoro, 2-bromo-5-fluorobenzenamine, 2-bromo-5-fluoro-aniline, 2-bromo-5-fluoro-phenylamine, 2-bromo-5-fluoro-benzenamine, 2-bromo-5-fluorophenylamine, pubchem1524, 5-fluoro-2-bromoaniline, 2-bromo-5-fluoro aniline, acmc-1c3a5
PubChem CID 2773317
IUPAC Name 2-bromo-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Br

Specifications

Melting Point 42°C to 46°C
Boiling Point 154°C (23 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN2811
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only