missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-chloroanisole 98.0+%, TCI America™
$59.69 - $194.44
Chemical Identifiers
| CAS | 174913-09-8 |
|---|---|
| Molecular Formula | C7H6BrClO |
| Molecular Weight (g/mol) | 221.478 |
| MDL Number | MFCD03790889 |
| InChI Key | CQGYLDZGJLVLMK-UHFFFAOYSA-N |
| Synonym | 2-bromo-5-chloroanisole, benzene, 1-bromo-4-chloro-2-methoxy, pubchem5265, acmc-1c20c, ksc497i0p, 2-bromo-5-chloroanisole, 98+%, 2-methoxy-4-chloro-1-bromobenzene, 1-bromo-4-chloro-2-methoxy-benzene |
| PubChem CID | 17984845 |
| IUPAC Name | 1-bromo-4-chloro-2-methoxybenzene |
| SMILES | COC1=C(C=CC(=C1)Cl)Br |
Chemical Identifiers
| 174913-09-8 | |
| 221.478 | |
| CQGYLDZGJLVLMK-UHFFFAOYSA-N | |
| 17984845 | |
| COC1=C(C=CC(=C1)Cl)Br |
| C7H6BrClO | |
| MFCD03790889 | |
| 2-bromo-5-chloroanisole, benzene, 1-bromo-4-chloro-2-methoxy, pubchem5265, acmc-1c20c, ksc497i0p, 2-bromo-5-chloroanisole, 98+%, 2-methoxy-4-chloro-1-bromobenzene, 1-bromo-4-chloro-2-methoxy-benzene | |
| 1-bromo-4-chloro-2-methoxybenzene |
Specifications
| 174913-09-8 | |
| C7H6BrClO | |
| 5 g | |
| CQGYLDZGJLVLMK-UHFFFAOYSA-N | |
| 1-bromo-4-chloro-2-methoxybenzene | |
| 17984845 | |
| ≥98.0% (GC) | |
| 2-Bromo-5-chloroanisole |
| Yellow | |
| MFCD03790889 | |
| 2-bromo-5-chloroanisole, benzene, 1-bromo-4-chloro-2-methoxy, pubchem5265, acmc-1c20c, ksc497i0p, 2-bromo-5-chloroanisole, 98+%, 2-methoxy-4-chloro-1-bromobenzene, 1-bromo-4-chloro-2-methoxy-benzene | |
| COC1=C(C=CC(=C1)Cl)Br | |
| 221.478 | |
| 221.48 | |
| Liquid |
Safety and Handling
TSCA : No