accessibility menu, dialog, popup

UserName

CAS RN 174913-09-8

IUPAC Name: 1-bromo-4-chloro-2-methoxybenzene
Synonyms: 1-bromo-4-chloro-2-methoxybenzene
Molecular Weight (g/mol): 221.48
Molecular Formula: C7H6BrClO
InChi Key: CQGYLDZGJLVLMK-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=CC(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)