missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chloroanisole 98.0+%, TCI America™

Supplier:  TCI America B364625G

 View more versions of this product

Catalog No. B364625G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Bromo-5-chloroanisole
Yellow
MFCD03790889
2-bromo-5-chloroanisole, benzene, 1-bromo-4-chloro-2-methoxy, pubchem5265, acmc-1c20c, ksc497i0p, 2-bromo-5-chloroanisole, 98+%, 2-methoxy-4-chloro-1-bromobenzene, 1-bromo-4-chloro-2-methoxy-benzene
COC1=C(C=CC(=C1)Cl)Br
221.478
221.48
Liquid
174913-09-8
C7H6BrClO
25 g
CQGYLDZGJLVLMK-UHFFFAOYSA-N
1-bromo-4-chloro-2-methoxybenzene
17984845
≥98.0% (GC)

Chemical Identifiers

174913-09-8
221.478
CQGYLDZGJLVLMK-UHFFFAOYSA-N
17984845
COC1=C(C=CC(=C1)Cl)Br
C7H6BrClO
MFCD03790889
2-bromo-5-chloroanisole, benzene, 1-bromo-4-chloro-2-methoxy, pubchem5265, acmc-1c20c, ksc497i0p, 2-bromo-5-chloroanisole, 98+%, 2-methoxy-4-chloro-1-bromobenzene, 1-bromo-4-chloro-2-methoxy-benzene
1-bromo-4-chloro-2-methoxybenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions