missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chloro-3-nitropyridine, 98%

Catalog No. AAH6406514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6406514 25 g
AAH6406506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6406514 Supplier Thermo Scientific Chemicals Supplier No. H6406514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75806-86-9 | C5H2BrClN2O2 | 237.437 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75806-86-9
Molecular Formula C5H2BrClN2O2
Molecular Weight (g/mol) 237.437
MDL Number MFCD07368982
InChI Key VFMIDVIAQMFGGP-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-5-chloropyridine, pyridine, 2-bromo-5-chloro-3-nitro, 2-bromo-5-chloro-3-nitro-pyridine, pubchem5951, acmc-1bggp, ksc495s5l, 2-bromo-5-chloro-3-nitro pyridine, 3-nitro-2-bromo-5-chloropyridine, 5-chloro-2-bromo-3-nitropyridine
PubChem CID 280681
IUPAC Name 2-bromo-5-chloro-3-nitropyridine
SMILES C1=C(C(=NC=C1Cl)Br)[N+](=O)[O-]

Specifications

Melting Point 175°C to 179°C
Color White
Quantity 25 g
UN Number UN2811
Beilstein 147540
Formula Weight 237.44
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-chloro-3-nitropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only