missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)pyridine, 97%

Catalog No. AAH2574806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2574806 5 g
AAH2574803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2574806 Supplier Thermo Scientific Chemicals Supplier No. H2574806
Only null left

CAS: 175205-81-9 | C6H3BrF3N | 225.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175205-81-9
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD00153085
InChI Key WZVHLUMAQLUNTJ-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethyl pyridine, pyridine, 2-bromo-4-trifluoromethyl, 2-bromo-4-trifluoromethyl-pyridine, 2-bromo-4-trifluormethyl pyridine, pubchem8064, acmc-209e9d, abbypharma ap-11-5529
PubChem CID 2781543
IUPAC Name 2-bromo-4-(trifluoromethyl)pyridine
SMILES C1=CN=C(C=C1C(F)(F)F)Br

Specifications

Density 1.83
Boiling Point 84°C to 85°C (14 mmHg)
Flash Point 73°C (164°F)
Refractive Index 1.478
Quantity 5 g
Formula Weight 226
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only