Learn More
2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%
CAS: 103962-10-3 | C9H6BrF3O2 | 283.044 g/mol
$120.92 - $430.74
Chemical Identifiers
| CAS | 103962-10-3 |
|---|---|
| Molecular Formula | C9H6BrF3O2 |
| Molecular Weight (g/mol) | 283.044 |
| MDL Number | MFCD00052333 |
| InChI Key | AOAGGWLQIILIIV-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl |
| PubChem CID | 2778691 |
| IUPAC Name | 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone |
| SMILES | C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6410503
|
Thermo Scientific Chemicals
H6410503 |
1 g |
Each for $120.92
|
|
|||||
|
AAH6410506
|
Thermo Scientific Chemicals
H6410506 |
5 g |
Each for $430.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 103962-10-3 | |
| 283.044 | |
| AOAGGWLQIILIIV-UHFFFAOYSA-N | |
| 2778691 | |
| C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F |
| C9H6BrF3O2 | |
| MFCD00052333 | |
| 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl | |
| 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone |
Specifications
| 103962-10-3 | |
| Yellow | |
| C9H6BrF3O2 | |
| 1 g | |
| 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl | |
| C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F | |
| 283.044 | |
| 283.04 | |
| Solid |
| 48°C to 55°C | |
| >110°C (230°F) | |
| MFCD00052333 | |
| 7479010 | |
| AOAGGWLQIILIIV-UHFFFAOYSA-N | |
| 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone | |
| 2778691 | |
| 97% | |
| 2-Bromo-4'-(trifluoromethoxy)acetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only