accessibility menu, dialog, popup

UserName

OBrOFFF

CAS RN 103962-10-3

IUPAC Name: 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 283.04
Molecular Formula: C9H6BrF3O2
InChi Key: AOAGGWLQIILIIV-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(C=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results