missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%

CAS: 103962-10-3 | C9H6BrF3O2 | 283.044 g/mol

Supplier:  Thermo Scientific Chemicals H6410506

 View more versions of this product

Catalog No. AAH6410506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-4'-(trifluoromethoxy)acetophenone
48°C to 55°C
>110°C (230°F)
MFCD00052333
7479010
AOAGGWLQIILIIV-UHFFFAOYSA-N
2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone
2778691
97%
103962-10-3
Yellow
C9H6BrF3O2
5 g
4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl
C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F
283.044
283.04
Solid

Chemical Identifiers

103962-10-3
283.044
AOAGGWLQIILIIV-UHFFFAOYSA-N
2778691
C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F
C9H6BrF3O2
MFCD00052333
4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl
2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only