Learn More
2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%
CAS: 103962-10-3 | C9H6BrF3O2 | 283.044 g/mol
Supplier: Thermo Scientific Chemicals H6410503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Bromo-4'-(trifluoromethoxy)acetophenone | |
| 48°C to 55°C | |
| >110°C (230°F) | |
| MFCD00052333 | |
| 7479010 | |
| AOAGGWLQIILIIV-UHFFFAOYSA-N | |
| 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone | |
| 2778691 | |
| 97% |
| 103962-10-3 | |
| Yellow | |
| C9H6BrF3O2 | |
| 1 g | |
| 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl | |
| C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F | |
| 283.044 | |
| 283.04 | |
| Solid |
Chemical Identifiers
| 103962-10-3 | |
| 283.044 | |
| AOAGGWLQIILIIV-UHFFFAOYSA-N | |
| 2778691 | |
| C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F |
| C9H6BrF3O2 | |
| MFCD00052333 | |
| 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl | |
| 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only