missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-tert-butylphenol 97.0+%, TCI America™

Catalog No. B37635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37635G 5 g
B376325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37635G Supplier TCI America Supplier No. B37635G
Only null left

Chemical Identifiers

CAS 2198-66-5
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.117
MDL Number MFCD02682891
InChI Key FFRLMQPMGIMHHQ-UHFFFAOYSA-N
PubChem CID 75147
IUPAC Name 2-bromo-4-tert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C=C1)O)Br

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 113°C
Quantity 5 g
Formula Weight 229.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-4-tert-butylphenol