missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitrotoluene 98.0+%, TCI America™

Catalog No. B126225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B126225G 25 g
B1262250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B126225G Supplier TCI America Supplier No. B126225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7745-93-9
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007195
InChI Key XFZFJQHXWJIBQV-UHFFFAOYSA-N
Synonym 2-bromo-4-nitrotoluene, benzene, 2-bromo-1-methyl-4-nitro, 3-bromo-4-methylnitrobenzene, toluene, 2-bromo-4-nitro, 3-bromo-4-methyl nitrobenzene, 1-bromo-2-methyl-5-nitrobenzene, 2-bromo-1-methyl-4-nitro-benzene, pubchem3822, 2-bromo-4nitrotoluene, 2-bromo4-nitrotoluene
PubChem CID 82189
IUPAC Name 2-bromo-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])Br

Specifications

Melting Point 77°C
Boiling Point 175°C
Quantity 25 g
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitrotoluene