missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-nitroacetophenone, 95%

Catalog No. AC305480500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305480500 50 g Glass bottle
AC305480100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC305480500 Supplier Thermo Scientific Chemicals Supplier No. 305480500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-81-0 | C8H6BrNO3 | 244.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-81-0
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.04
MDL Number MFCD00007356
InChI Key MBUPVGIGAMCMBT-UHFFFAOYSA-N
Synonym 2-bromo-4'-nitroacetophenone, 2-bromo-1-4-nitrophenyl ethanone, 4-nitrophenacyl bromide, p-nitrophenacyl bromide, 2-bromo-1-4-nitrophenyl ethan-1-one, alpha-bromo-p-nitroacetophenone, ethanone, 2-bromo-1-4-nitrophenyl, 4'-nitrophenacyl bromide, alpha-bromo-4-nitroacetophenone, 2-bromo-4-nitroacetophenone
PubChem CID 66840
IUPAC Name 2-bromo-1-(4-nitrophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)[N+](=O)[O-]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 94°C to 100°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COCH2Br
Quantity 50 g
Formula Weight 244.04
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-nitroacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-789-9

RUO – Research Use Only