missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-fluoroaniline 98.0+%, TCI America™
Supplier: TCI America B182925G
Specifications
| 2-Bromo-4-fluoroaniline | |
| 221°C | |
| MFCD00042462 | |
| 2-bromo-4-fluorobenzenamine, 4-fluoro-2-bromoaniline, 2-bromo-4-fluoro-aniline, benzenamine, 2-bromo-4-fluoro, 2-bromo-4-fluoro-phenylamine, 2-bromo-4-fluoro aniline, 2-bromo-4-fluorophenylamine, 2-brom-4-fluoranilin, bromofluoroaniline1, pubchem2918 | |
| C1=CC(=C(C=C1F)Br)N | |
| 190.015 | |
| 190.02 | |
| Crystalline Powder |
| 1003-98-1 | |
| C6H5BrFN | |
| 25 g | |
| YLMFXCIATJJKQL-UHFFFAOYSA-N | |
| 2-bromo-4-fluoroaniline | |
| 242873 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1003-98-1 | |
| 190.015 | |
| YLMFXCIATJJKQL-UHFFFAOYSA-N | |
| 242873 | |
| C1=CC(=C(C=C1F)Br)N |
| C6H5BrFN | |
| MFCD00042462 | |
| 2-bromo-4-fluorobenzenamine, 4-fluoro-2-bromoaniline, 2-bromo-4-fluoro-aniline, benzenamine, 2-bromo-4-fluoro, 2-bromo-4-fluoro-phenylamine, 2-bromo-4-fluoro aniline, 2-bromo-4-fluorophenylamine, 2-brom-4-fluoranilin, bromofluoroaniline1, pubchem2918 | |
| 2-bromo-4-fluoroaniline |
Safety and Handling
TSCA : No