missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoroaniline 98.0+%, TCI America™

Catalog No. p-7119289
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18295G 5 g
B182925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18295G Supplier TCI America Supplier No. B18295G
Only null left

Chemical Identifiers

CAS 1003-98-1
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD00042462
InChI Key YLMFXCIATJJKQL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzenamine, 4-fluoro-2-bromoaniline, 2-bromo-4-fluoro-aniline, benzenamine, 2-bromo-4-fluoro, 2-bromo-4-fluoro-phenylamine, 2-bromo-4-fluoro aniline, 2-bromo-4-fluorophenylamine, 2-brom-4-fluoranilin, bromofluoroaniline1, pubchem2918
PubChem CID 242873
IUPAC Name 2-bromo-4-fluoroaniline
SMILES C1=CC(=C(C=C1F)Br)N

Specifications

Boiling Point 221°C
Quantity 5 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-fluoroaniline