accessibility menu, dialog, popup

UserName

CAS RN 1003-98-1

IUPAC Name: 2-bromo-4-fluoroaniline
Synonyms: 2-bromo-4-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: YLMFXCIATJJKQL-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C=C1Br
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)