missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-6-(trifluoromethyl)aniline, 98%

Catalog No. AAH2620609
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2620609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2620609 Supplier Thermo Scientific Chemicals Supplier No. H2620609

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 875664-27-0 | C7H4BrF4N | 258.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 875664-27-0
Molecular Formula C7H4BrF4N
Molecular Weight (g/mol) 258.014
MDL Number MFCD04973756
InChI Key OTYCOVZULSPPPW-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-6-trifluoromethyl aniline, 2-amino-3-bromo-5-fluorobenzotrifluoride, buttpark 154\11-38, benzenamine,2-bromo-4-fluoro-6-trifluoromethyl, 2-bromo-4-fluoro-6-trifluoromethyl benzenamine, pubchem19869, ksc658m8n, 2-bromo-4-fluoro-6-trifluoromethyl phenylamine, 6-bromo-4-fluoro-2-trifluoromethyl phenylamine, 2-bromanyl-4-fluoranyl-6-trifluoromethyl aniline
PubChem CID 7018045
IUPAC Name 2-bromo-4-fluoro-6-(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1Br)N)C(F)(F)F)F

Specifications

Refractive Index 1.503
Quantity 10 g
UN Number UN2810
Formula Weight 258.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluoro-6-(trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only