missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chlorophenol, 98+%

Catalog No. AAA1621822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1621822 100 g
AAA1621814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1621822 Supplier Thermo Scientific Chemicals Supplier No. A1621822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 695-96-5 | C6H4BrClO | 207.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 695-96-5
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.45
MDL Number MFCD00002319
InChI Key ZIYRDJLAJYTELF-UHFFFAOYSA-N
Synonym phenol, 2-bromo-4-chloro, 4-chloro-2-bromophenol, 2-bromo-4-chloro-phenol, 4-chloro-2-bromo phenol, pubchem3594, 2-bromo-4-chorophenol, acmc-1ayt0, akos bbb/602, ksc493o9n, 2-bromo-4-chlorophenol
PubChem CID 69670
IUPAC Name 2-bromo-4-chlorophenol
SMILES OC1=CC=C(Cl)C=C1Br

Specifications

Melting Point 31°C to 35°C
Boiling Point 230°C to 232°C
Flash Point >110°C (230°F)
Odor Phenol-like
Quantity 100 g
Beilstein 2042871
Formula Weight 207.46
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4-chlorophenol
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-785-6
TSCA No

RUO – Research Use Only