missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloro-1-iodobenzene 96.0+%, TCI America™

Catalog No. p-7123426
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42435G 5 g
B424325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42435G Supplier TCI America Supplier No. B42435G

Chemical Identifiers

CAS 31928-44-6
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.348
MDL Number MFCD00672944
InChI Key CXHXFDQEFKFYQJ-UHFFFAOYSA-N
Synonym 2-bromo-4-chloroiodobenzene, benzene, 2-bromo-4-chloro-1-iodo, acmc-209hpo, ksc494q6f, 1-iodo-2-bromo-4-chlorobenzene, 2-bromo-1-iodo-4-chlorobenzene
PubChem CID 13741326
IUPAC Name 2-bromo-4-chloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Br)I

Specifications

Melting Point 33°C
Color Yellow
Boiling Point 111°C
Quantity 5 g
Formula Weight 317.35
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-4-chloro-1-iodobenzene