missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dimethylpyridine 98.0+%, TCI America™

Catalog No. p-7124026
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52541G 1 g
B52545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52541G Supplier TCI America Supplier No. B52541G

Chemical Identifiers

CAS 4926-26-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00082591
InChI Key IRTOCXBLUOPRFT-UHFFFAOYSA-N
Synonym 2-Bromo-4,6-lutidine
PubChem CID 14975195
IUPAC Name 2-bromo-4,6-dimethylpyridine
SMILES CC1=CC(C)=NC(Br)=C1

Specifications

Color Yellow
Boiling Point 68°C
Quantity 1 g
Formula Weight 186.05
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4,6-dimethylpyridine