missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7119419
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20115G 5 g
B201125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20115G Supplier TCI America Supplier No. B20115G

Chemical Identifiers

CAS 5392-10-9
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00003301
InChI Key UQQROBHFUDBOOK-UHFFFAOYSA-N
Synonym 6-bromoveratraldehyde, 6-bromo-3,4-dimethoxybenzaldehyde, benzaldehyde, 2-bromo-4,5-dimethoxy, 6-bromo veratraldehyde, 2-bromo-4,5-dimethoxybenzalehyde, 4,5-dimethoxy-2-bromobenzaldehyde, 6-bromoveratral, pubchem8209, veratraldehyde, 6-bromo, acmc-2097hh
PubChem CID 79351
IUPAC Name 2-bromo-4,5-dimethoxybenzaldehyde
SMILES COC1=C(C=C(C(=C1)C=O)Br)OC

Specifications

Melting Point 148°C
Quantity 5 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,5-dimethoxybenzaldehyde