missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorotoluene 98.0+%, TCI America™

Catalog No. B41295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41295G 5 g
B41291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41295G Supplier TCI America Supplier No. B41295G
Only null left

Chemical Identifiers

CAS 875664-38-3
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD07777160
InChI Key FKEURBCLFHOBDM-UHFFFAOYSA-N
Synonym 2-bromo-4,5-difluorotoluene, 1-bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methylbromobenzene, ksc495c2l, bromo-4,5-difluoro-2-methyl-benzene, 4,5-difluoro-2-methyl-1-bromobenzene, 1-brommo-4,5-difluoro-2-methyl-benzene, benzene,1-bromo-4,5-difluoro-2-methyl
PubChem CID 44890785
IUPAC Name 1-bromo-4,5-difluoro-2-methylbenzene
SMILES CC1=CC(=C(C=C1Br)F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 207.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4,5-difluorotoluene