missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(2-thienyl)thiazole, 97%, Thermo Scientific™

Catalog No. AAH5857504
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5857504 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5857504 Supplier Thermo Scientific Chemicals Supplier No. H5857504
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1125409-90-6
Molecular Formula C7H4BrNS2
Molecular Weight (g/mol) 246.14
MDL Number MFCD11220135
InChI Key XRXGEQOSIYEEKD-UHFFFAOYSA-N
Synonym 2-bromo-4-2-thienyl thiazole, 2-bromo-4-thiophen-2-yl-1,3-thiazole
PubChem CID 38996210
IUPAC Name 2-bromo-4-thiophen-2-yl-1,3-thiazole
SMILES C1=CSC(=C1)C2=CSC(=N2)Br

Specifications

Quantity 2 g
Beilstein 21223492
Formula Weight 246.14
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(2-thienyl)thiazole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures