missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-(1-pyrrolidinyl)acetophenone, 97%

Catalog No. AAL1540714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1540714 25 g
AAL1540706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1540714 Supplier Thermo Scientific Chemicals Supplier No. L1540714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 216144-18-2 | C12H14BrNO | 268.154 g/mol

2-Bromo-4'-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-(1-pyrrolidinyl)acetophenone is an important raw material and used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Chemical Identifiers

CAS 216144-18-2
Molecular Formula C12H14BrNO
Molecular Weight (g/mol) 268.154
MDL Number MFCD01075690
InChI Key WLRIUCQUMKJOGT-UHFFFAOYSA-N
Synonym 2-bromo-1-4-pyrrolidin-1-yl phenyl ethanone, alpha-bromo-4-1-pyrrolidino acetophenone, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl ethanone, ethanone, 2-bromo-1-4-1-pyrrolidinyl phenyl, 2-bromo-1-4-pyrrolidin-1-yl phenyl ethan-1-one, 2-bromo-1-4-pyrrolidin-1-ylphenyl ethan-1-one, 4-pyrrolidin-1-yl phenacyl bromide, 2-bromo-1-4-pyrrolidinylphenyl ethan-1-one, acmc-209fm8
PubChem CID 3681785
IUPAC Name 2-bromo-1-(4-pyrrolidin-1-ylphenyl)ethanone
SMILES C1CCN(C1)C2=CC=C(C=C2)C(=O)CBr

Specifications

Melting Point 120°C to 122°C
Quantity 25 g
UN Number UN3261
Beilstein 8549816
Solubility Information Insoluble in water.
Formula Weight 268.16
Percent Purity 97%
Chemical Name or Material 2-Bromo-4'-(1-pyrrolidinyl)acetophenone
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only