missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-fluoroaniline, 97%

Catalog No. AAH6168706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6168706 5 g
AAH6168703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6168706 Supplier Thermo Scientific Chemicals Supplier No. H6168706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111721-75-6 | C6H5BrFN | 190.015 g/mol

2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-3-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 111721-75-6
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD07369915
InChI Key XZRSXRUYZXBTGD-UHFFFAOYSA-N
Synonym 2-bromo-3-fluorophenylamine, 2-bromo-3-fluoro-phenylamine, 2-bromo-3-fluorobenzenamine, benzenamine, 2-bromo-3-fluoro, pubchem1523, acmc-1c6kb, 2-bromo-3-fluoro-aniline, ksc493i1t, 2-bromo-3-fluoro-phenyl-amine
PubChem CID 14770005
IUPAC Name 2-bromo-3-fluoroaniline
SMILES C1=CC(=C(C(=C1)F)Br)N

Specifications

Color White
Density 1.670 g/mL
Flash Point 94°C (201°F)
Refractive Index 1.583
Quantity 5 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 190.01
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 2-Bromo-3-fluoroaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only