missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3205506

 View more versions of this product

Catalog No. AAH3205506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
80°C (32 mmHg)
MFCD00153072
2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine
C1=C(C=NC(=C1Cl)Br)C(F)(F)F
260.438
260.44
75806-84-7
C6H2BrClF3N
5 g
SMTKGMYGLYWNDL-UHFFFAOYSA-N
2-bromo-3-chloro-5-(trifluoromethyl)pyridine
2736237
97%

Chemical Identifiers

75806-84-7
260.438
SMTKGMYGLYWNDL-UHFFFAOYSA-N
2736237
C1=C(C=NC(=C1Cl)Br)C(F)(F)F
C6H2BrClF3N
MFCD00153072
2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine
2-bromo-3-chloro-5-(trifluoromethyl)pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only